Tisk…

3-Bromo-2-nitropyridine ≥98%

Dodavatel: Thermo Scientific
Varování

L19523.06 L19523.03
L19523.06EA 5700 CZK
L19523.06 L19523.03
3-Bromo-2-nitropyridine ≥98%
3-Bromo-2-nitropyridine

Purity: 98+% CAS No.: 54231-33-3 Molecular Formula: C5H3BrN2O2 Formula Weight: 203.00 Melting Point: 102-106° Beilstein Reference: 1637071

Vzorec: C₅H₃BrN₂O₂
MW: 203 g/mol
Bod tání: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00955614
CAS číslo: 54231-33-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR