2,6-Dibromopyridine, Sigma-Aldrich®
Dodavatel: Merck
D43115-100G
D43115-25G
SIALD43115-100GEA
7470
CZK
SIALD43115-100G
SIALD43115-25G
2,6-Dibromopyridine, Sigma-Aldrich®
2,6-Dibromopyridine
Vzorec:
C₅H₃Br₂N MW: 236.89 Bod varu: 255 Bod tání: 116…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006223 CAS číslo: 626-05-1 EINECS: 210-926-9 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...