Tisk…

Pyridazine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

P57204-50G P57204-5G
SIALP57204-50GEA 12140 CZK
SIALP57204-50G SIALP57204-5G
Pyridazine, Sigma-Aldrich®
Pyridazine
Vzorec: C₄H₄N₂
MW: 80,09 g/mol
Bod varu: 208 °C (1013 hPa)
Bod tání: 0 °C
Hustota: 1,103 g/cm³ (20 °C)
MDL Number: MFCD00006463
CAS číslo: 289-80-5
EINECS: 206-025-5
Merck Index: 12,08152

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR