Tisk…

Pyridazine ≥98%

Dodavatel: Thermo Scientific
Varování

A13266.06 A13266.14
A13266.06EA 1520 CZK
A13266.06 A13266.14
Pyridazine ≥98%
Pyridazine

Purity: 98+% CAS No.: 289-80-5 Molecular Formula: C4H4N2 Formula Weight: 80.09 Flash Point: 85°(185°F) Refractive Index: 1.5240 Odour: Unpleasant Melting Point: -8° Boiling Point: 87°/14mm Solubility: Miscible with water, benzene, DMF. Soluble in alcohol, ether Beilstein Reference: 103906 Merck Reference: 14,7969 MDL No.: MFCD00006463

Vzorec: C₄H₄N₂
MW: 80,09 g/mol
Bod varu: 87 °C (14 mmHg)
Bod tání: –8 °C
Hustota: 1,105
Bod vzplanutí: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006463
CAS číslo: 289-80-5
EINECS: 206-025-5
Merck Index: 14,7969

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR