3-Chloro-6-methoxypyridazine ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
A14173.06
A14173.14
A14173.22
A14173.06EA
1390
CZK
A14173.06
A14173.14
A14173.22
3-Chloro-6-methoxypyridazine ≥97%
3-Chloro-6-methoxypyridazine
Purity: 97%
CAS No.: 1722-10-7
Molecular Formula: C5H5ClN2O
Formula Weight: 144.56
Melting Point: 90-92°
Beilstein Reference: 118854
Vzorec:
C₅H₅ClN₂O MW: 144,56 g/mol Bod varu: 174…175 °C (3 mmHg) Bod tání: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006467 CAS číslo: 1722-10-7 EINECS: 217-019-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...