3,5-Dimethyl-1H-pyrazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3,5-Dimethylpyrazol, 3,5-Dimethyl-1H-pyrazole, 3,5-Dimethylpyrazole
M-Clarity™ quality level = MQ100
05091-100G-F
05091-1KG-F
05091-25KG-F
SIAL05091-100G-FEA
1980
CZK
SIAL05091-100G-F
SIAL05091-1KG-F
SIAL05091-25KG-F
3,5-Dimethyl-1H-pyrazole, Sigma-Aldrich®
3,5-Dimethyl-1H-pyrazole
Vzorec:
C₅H₈N₂ MW: 96,13 g/mol Bod varu: 218 °C (1013 hPa) Hustota: 0,8839 g/cm³ (16 °C) |
MDL Number:
MFCD00005243 CAS číslo: 67-51-6 EINECS: 200-657-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...