Tisk…

2,3,5,6-Tetramethylpyrazine ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A18279.14 A18279.22
A18279.14EA 1650 CZK
A18279.14 A18279.22
2,3,5,6-Tetramethylpyrazine ≥98%
2,3,5,6-Tetramethylpyrazine

Purity: 98+% CAS No.: 1124-11-4 Molecular Formula: C8H12N2 Formula Weight: 136.20 Odour: Pleasant pH: 7 Melting Point: 83-86° Boiling Point: 190° Beilstein Reference: 113100 MDL No.: MFCD00006146

Vzorec: C₈H₁₂N₂
MW: 136,2 g/mol
Bod varu: 190 °C
Bod tání: 83…86 °C
Hustota: 1,08
Storage Temperature: Ambient
MDL Number: MFCD00006146
CAS číslo: 1124-11-4
EINECS: 214-391-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR