2-Ethyl-3-methylpyrazine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Purity: 98+%
CAS No.: 15707-23-0
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 57°(134°F)
Refractive Index: 1.5030
Odour: Characteristic
Boiling Point: 169-171°
Beilstein Reference: 956775
UN No.: 1993
MDL No.: MFCD00006150
Vzorec:
C₇H₁₀N₂ Bod varu: 169…171 °C Hustota: 0,987 Bod vzplanutí: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006150 CAS číslo: 15707-23-0 EINECS: 239-799-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...