2,6-Dimethylpyrazine ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Purity: 99%
CAS No.: 108-50-9
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 52°(125°F)
Refractive Index: 1.5000
Odour: Aromatic
Melting Point: 38-43°
Boiling Point: 153-154°
Beilstein Reference: 1726
UN No.: 1325
MDL No.: MFCD00006148
Vzorec:
C₆H₈N₂ Bod varu: 153…154 °C Bod tání: 38…43 °C Hustota: 0,9647 g/cm³ (50 °C) Bod vzplanutí: 52 °C (125 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006148 CAS číslo: 108-50-9 EINECS: 203-589-4 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...