2,5-Dimethylpyrazine ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
A12352.06
A12352.14
A12352.22
A12352.06EA
1670
CZK
A12352.06
A12352.14
A12352.22
2,5-Dimethylpyrazine ≥99%
2,5-Dimethylpyrazine
Purity: 99%
CAS No.: 123-32-0
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 63°(145°F)
Refractive Index: 1.5 00
Odour: Pleasant
Melting Point: 15°
Boiling Point: 154-155°
Beilstein Reference: 107052
MDL No.: MFCD00006147
Vzorec:
C₆H₈N₂ MW: 108,14 g/mol Bod varu: 154…155 °C Bod tání: 15 °C Hustota: 0,990 Bod vzplanutí: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006147 CAS číslo: 123-32-0 EINECS: 204-618-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...