Tisk…

2-Methylpyrazine ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

A14146.14 A14146.22 A14146.36
A14146.14EA 1140 CZK
A14146.14 A14146.22 A14146.36
2-Methylpyrazine ≥99%
2-Methylpyrazine

Purity: 99+% CAS No.: 109-08-0 Molecular Formula: C5H6N2 Formula Weight: 94.12 Flash Point: 50°(122°F) Refractive Index: 1.5050 Odour: Unpleasant Melting Point: -30° Boiling Point: 135-136° Beilstein Reference: 105778 UN No.: 1993 MDL No.: MFCD00006142

Vzorec: C₅H₆N₂
MW: 94,12 g/mol
Bod varu: 135…136 °C
Bod tání: –30 °C
Hustota: 1,028
Bod vzplanutí: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006142
CAS číslo: 109-08-0
EINECS: 203-645-8
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR