Pyrazine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
P56003-10G
P56003-25G
P56003-5G
SIALP56003-10GEA
1330
CZK
SIALP56003-10G
SIALP56003-25G
SIALP56003-5G
Pyrazine, Sigma-Aldrich®
Pyrazine
Vzorec:
C₄H₄N₂ MW: 80,09 g/mol Bod varu: 115 °C (1013 hPa) Bod tání: 52 °C Hustota: 1,0311 g/cm³ (60.9 °C) |
MDL Number:
MFCD00006122 CAS číslo: 290-37-9 EINECS: 206-027-6 UN: 1325 ADR: 4.1,III Merck Index: 13,08047 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...