Tisk…

β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD

M-Clarity™ quality level = MQ200

N7381-12G N7381-12X600MG N7381-20G
SIALN7381-12GEA 21780 CZK
SIALN7381-12G SIALN7381-12X600MG SIALN7381-20G
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate
Vzorec: C₂₁H₂₇N₇O₁₄P₂·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00036253
CAS číslo: 53-84-9
EINECS: 200-184-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR