Tisk…

β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD

M-Clarity™ quality level = MQ200

N1511-5G N1511-250MG N1511-1G
SIALN1511-5GEA 35230 CZK
SIALN1511-5G SIALN1511-250MG SIALN1511-1G
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate
Vzorec: C₂₁H₂₇N₇O₁₄P₂·H₂O
MDL Number: MFCD00036253
CAS číslo: 53-84-9
EINECS: 200-184-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR