Tisk…

3-Methylxanthine ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine

L14986.03 L14986.MD
L14986.03EA 8450 CZK
L14986.03 L14986.MD
3-Methylxanthine ≥98%
3-Methylxanthine

Purity: 98+% CAS No.: 1076-22-8 Molecular Formula: C6H6N4O2 Formula Weight: 166.14 Melting Point: 300° MDL No.: MFCD00005580

Vzorec: C₆H₆N₄O₂
MW: 166,14 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005580
CAS číslo: 1076-22-8
EINECS: 214-058-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR