Guanine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
G11950-25G
G11950-100G
G11950-10G
SIALG11950-25GEA
2090
CZK
SIALG11950-25G
SIALG11950-100G
SIALG11950-10G
Guanine, Sigma-Aldrich®
Guanine
Vzorec:
C₅H₅N₅O Bod tání: > 220 °C Hustota: 1,67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071533 CAS číslo: 73-40-5 EINECS: 200-799-8 Merck Index: 13,04580 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...