Adenin, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine
M-Clarity™ quality level = MQ200
A5665-100G
A5665-25G
SIALA5665-100GEA
11720
CZK
SIALA5665-100G
SIALA5665-25G
Adenin, Sigma-Aldrich®
Adenin
Vzorec:
C₅H₅N₅ MW: 135,13 g/mol Hustota: 0,99741 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00041790 CAS číslo: 73-24-5 EINECS: 200-796-1 UN: 2811 ADR: 6.1,III Merck Index: 13,00152 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...