Tisk…

Adenin, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

M-Clarity™ quality level = MQ200

A2786-25G A2786-100G A2786-5G
SIALA2786-25GEA 3980 CZK
SIALA2786-25G SIALA2786-100G SIALA2786-5G
Adenin, Sigma-Aldrich®
Adenin
Vzorec: C₅H₅N₅
MW: 135,13 g/mol
Hustota: 0,99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS číslo: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00152

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR