Tisk…

2-Amino-6-chloropurine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine

M-Clarity™ quality level = MQ200

109789-1G 109789-5G
SIAL109789-1GEA 2250 CZK
SIAL109789-1G SIAL109789-5G
2-Amino-6-chloropurine, Sigma-Aldrich®
2-Amino-6-chloropurine
Vzorec: C₅H₄ClN₅
MW: 169,57 g/mol
Bod tání: 300 °C
MDL Number: MFCD00075252
CAS číslo: 10310-21-1
EINECS: 233-686-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR