Tisk…

2,6-Dichloropurine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,6-Dichloro-9H-purine

M-Clarity™ quality level = MQ100

D73103-10G D73103-1G D73103-5G
SIALD73103-10GEA 0 CZK
SIALD73103-10G SIALD73103-1G SIALD73103-5G
2,6-Dichloropurine, Sigma-Aldrich®
2,6-Dichloropurine
Vzorec: C₅H₂Cl₂N₄
MW: 189 g/mol
Bod tání: 181 °C
MDL Number: MFCD00077725
CAS číslo: 5451-40-1
EINECS: 226-681-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR