Tisk…

2,6-Dichloropurine ≥97%

Dodavatel: Thermo Scientific

Synonyms: 2,6-Dichloro-9H-purine

B21289.03 B21289.06
B21289.03EA 1470 CZK
B21289.03 B21289.06
2,6-Dichloropurine ≥97%
2,6-Dichloropurine

Purity: 97% CAS No.: 5451-40-1 Molecular Formula: C5H2Cl2N4 Formula Weight: 189.01 Melting Point: ca 184-186° dec. Beilstein Reference: 9354

Vzorec: C₅H₂Cl₂N₄
MW: 189 g/mol
Bod tání: ∼ 184…186 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00077725
CAS číslo: 5451-40-1
EINECS: 226-681-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR