Tisk…
Pluronic® F-127, powder, BioReagent cell culture reagent, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: PEG-PPG-PEG, Pluronic® F-127
M-Clarity™ quality level = MQ200
P2443-1KG P2443-250G
SIALP2443-1KGEA 13180 CZK
SIALP2443-1KG SIALP2443-250G
Pluronic® F-127, powder, BioReagent cell culture reagent, Sigma-Aldrich®
Poloxamer 407 (Pluronic® F-127)
A non-ionic copolymer surfactant qualified for use in insect cell culture applications as an antifoaming agent.
  • Suitable for cell culture
  • CMC: 950 to 1000 ppm (~25 °C)
Other Applications: Pluronic® F-127 was used to coat a siliconised coverslip to hold an egg extract in a study. Pluronic F-127 was added to phosphate buffered saline (PBS), to lower unspecific cell and protein adhesion to a PDMS-based microfluidic device. A study reports its use as a release vehicle to transport low-dose perivascular lipopolysaccharide (LPS) on mouse vein grafts. PLGA/pluronic F127 may be used to fabricate nerve guidance conduits (NGCs) for regeneration of peripheral nerve. Fabrication of poly (lactide-co-glycolide) (PLGA)-Pluronic F127 glass composites was reported. Pluronic F-127 was used for fluorescent labeling of blood vessels, astrocytes, and neurons.
Balení: Packed in poly bottle.
Upozornení: Pluronic is a registered trademark of BASF.
Vzorec: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number: MFCD00082049
CAS číslo: 9003-11-6