Tisk…

Phenazine ≥99%

Dodavatel: Thermo Fisher Scientific

A15770.06 A15770.14 A15770.22
A15770.06EA 1420 CZK
A15770.06 A15770.14 A15770.22
Phenazine ≥99%
Phenazine

Purity: 99+% CAS No.: 92-82-0 Molecular Formula: C12H8N2 Formula Weight: 180.21 Melting Point: 175-177° MDL No.: MFCD00005023

Vzorec: C₁₂H₈N₂
Bod varu: 339 °C (1013 hPa)
Bod tání: 175…177 °C
Hustota: 1,26 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005023
CAS číslo: 92-82-0
EINECS: 202-193-9
Merck Index: 12,07359

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR