Phenazine 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
130150050
130150100
ACRO130150050EA
1460
CZK
ACRO130150050
ACRO130150100
Phenazine 98%
Phenazine
Vzorec:
C₁₂H₈N₂ Bod varu: >360 °C (760 mmHg) Bod tání: 174…177 °C Hustota: 1,26 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005023 CAS číslo: 92-82-0 EINECS: 202-193-9 Merck Index: 12,07359 |
Výsledky specifikačního testu
Appearance | Yellow to brown Crystalline powder or crystals or needles |
Infrared spectrum | Conforms |
Melting point | 173 to 177 °C |
HPLC | ≥97.5 % |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.5 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...