9-Phenanthrenecarbaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
P11603-10G
P11603-2.5G
SIALP11603-10GEA
9270
CZK
SIALP11603-10G
SIALP11603-2.5G
9-Phenanthrenecarbaldehyde, Sigma-Aldrich®
9-Phenanthrenecarbaldehyde
Vzorec:
C₁₅H₁₀O Storage Temperature: Ambient |
MDL Number:
MFCD00001175 CAS číslo: 4707-71-5 EINECS: 225-194-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...