9-Phenanthrenecarbaldehyde ≥97%
Dodavatel: Thermo Fisher Scientific
B25648.03
B25648.06
B25648.03EA
1120
CZK
B25648.03
B25648.06
9-Phenanthrenecarbaldehyde ≥97%
9-Phenanthrenecarbaldehyde
Purity: 97%
CAS No.: 4707-71-5
Molecular Formula: C15H10O
Formula Weight: 206.24
Melting Point: 100-103°
MDL No.: MFCD00001175
Vzorec:
C₁₅H₁₀O Bod tání: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001175 CAS číslo: 4707-71-5 EINECS: 225-194-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...