L(+)-Kyselina askorbová sodná sůl, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
M-Clarity™ quality level = MQ200
A4034-100G
A4034-1KG
A4034-500G
SIALA4034-100GEA
2380
CZK
SIALA4034-100G
SIALA4034-1KG
SIALA4034-500G
L(+)-Kyselina askorbová sodná sůl, Sigma-Aldrich®
L(+)-Kyselina askorbová sodná sůl
Vzorec:
C₆H₇NaO₆ MW: 198.11 Bod tání: 219 Hustota: 1.799 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00082340 CAS číslo: 134-03-2 EINECS: 205-126-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...