Tisk…

Span® 85, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Arlacel 85, Sorbitan trioleate

M-Clarity™ quality level = MQ200

S7135-1L S7135-250ML
SIALS7135-1LEA 8360 CZK
SIALS7135-1L SIALS7135-250ML
Span® 85, Sigma-Aldrich®
Span® 85
Vzorec: C₆₀H₁₀₈O₈
MW: 957.51
Bod varu: ∼100 °C (1013 hPa)
Hustota: 0.95 g/cm³ (20 °C)
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00133820
CAS číslo: 26266-58-0
EINECS: 247-569-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR