Tisk…

Sorbitan monostearate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W302805-10KG W302805-1KG W302805-SAMPLE
SIALW302805-10KGEA 26990 CZK
SIALW302805-10KG SIALW302805-1KG SIALW302805-SAMPLE
Sorbitan monostearate, Sigma-Aldrich®
Sorbitan monostearate
Vzorec: C₂₄H₄₆O₆
Storage Temperature: Ambient
MDL Number: MFCD00005366
CAS číslo: 1338-41-6
EINECS: 215-664-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR