Tisk…

β-Butyrolactone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: beta-Methylpropiolactone, beta-Butyrolactone, 1,3-Butyrolactone, 4-Methyl-2-oxetanone, 4-Methyloxetan-2-one, Butan-3-olide

M-Clarity™ quality level = MQ100

219126-50G 219126-10G
SIAL219126-50GEA 2010 CZK
SIAL219126-50G SIAL219126-10G
β-Butyrolactone, Sigma-Aldrich®
β-Butyrolactone
Vzorec: C₄H₆O₂
MW: 86.09
Bod varu: 163
Bod tání: –43.5 °C
Hustota: 1.0555 g/cm³ (20 °C)
MDL Number: MFCD00005170
CAS číslo: 3068-88-0
EINECS: 221-330-3
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR