Tetrahydropyran, Sigma-Aldrich®
Dodavatel: Merck
MQ100
293105-100ML
293105-1L
SIAL293105-100MLEA
5310
CZK
SIAL293105-100ML
SIAL293105-1L
Tetrahydropyran, Sigma-Aldrich®
Tetrahydropyran
Vzorec:
C₅H₁₀O MW: 86.13 Bod varu: 88 Bod tání: –44.2 °C Hustota: 0.881 g/cm³ (20 °C) |
MDL Number:
MFCD00006585 CAS číslo: 142-68-7 EINECS: 205-552-8 UN: 3271 ADR: 3 II Merck Index: 12,09356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...