Tetrahydropyran 98
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13392.AE
A13392.AP
A13392.AEEA
1600
CZK
A13392.AE
A13392.AP
Tetrahydropyran 98
Tetrahydropyran
Purity: 98+%
CAS No.: 142-68-7
Molecular Formula: C5H10O
Formula Weight: 86.14
Flash Point: -15°(5°F)
Refractive Index: 1.4210
Melting Point: -49°
Boiling Point: 87-89°
Beilstein Reference: 102436
Merck Reference: 14,9217
UN No.: 3271
MDL No.: MFCD00006585
Vzorec:
C₅H₁₀O MW: 86.13 Bod varu: 87…89 °C Bod tání: –49 °C Hustota: 0.882 Bod vzplanutí: –15 °C (5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006585 CAS číslo: 142-68-7 EINECS: 205-552-8 UN: 3271 ADR: 3 II Merck Index: 12,09356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...