Tisk…

Pentadecan-15-olide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cyclopentadecanolide, Oxacyclohexadecan-2-one, Pentadecanolide, ω-Pentadecalactone, omega-Pentadecalactone, Oxa-2-cyclohexadecanone

M-Clarity™ quality level = MQ300

W284009-1KG-K W284009-5KG-K
SIALW284009-1KG-KEA 8790 CZK
SIALW284009-1KG-K SIALW284009-5KG-K
Pentadecan-15-olide, Sigma-Aldrich®
Pentadecan-15-olide
Vzorec: C₁₅H₂₈O₂
MDL Number: MFCD00039667
CAS číslo: 106-02-5
EINECS: 203-354-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR