Pentadecan-15-olide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Cyclopentadecanolide, Oxacyclohexadecan-2-one, Pentadecanolide, ω-Pentadecalactone, omega-Pentadecalactone, Oxa-2-cyclohexadecanone
M-Clarity™ quality level = MQ100
419133-100G
419133-25G
SIAL419133-100GEA
4690
CZK
SIAL419133-100G
SIAL419133-25G
Pentadecan-15-olide, Sigma-Aldrich®
Pentadecan-15-olide
Vzorec:
C₁₅H₂₈O₂ |
MDL Number:
MFCD00039667 CAS číslo: 106-02-5 EINECS: 203-354-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...