Kojic acid, white crystalline powder
Dodavatel: MP Biomedicals
Synonyms:
5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone
0215515710
0215515725
0215515701
0215515705
ICNA0215515710EA
7050
CZK
ICNA0215515710
ICNA0215515725
ICNA0215515701
ICNA0215515705
Kojic acid, white crystalline powder
Kojic acid
Kojic acid is used as an antibiotic reagent. It is tyrosinase inhibitor. Kojic acid is a chelation agent.
Soluble in water, ethanol, acetone; Slightly soluble in ethyl acetate, chloroform, pyridine.
Vzorec:
C₆H₆O₄ MW: 142,11 g/mol Bod varu: 323 °C (1013 hPa) Bod tání: 148…157 °C Hustota: 1,58 g/cm³ (20 °C) |
MDL Number:
MFCD00006580 CAS číslo: 501-30-4 EINECS: 207-922-4 Merck Index: 13,05334 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...