Tisk…

Kojic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A10760.06 A10760.14 A10760.22
A10760.06EA 1500 CZK
A10760.06 A10760.14 A10760.22
Kojic acid ≥99%
Kojic acid

Purity: 99% CAS No.: 501-30-4 Molecular Formula: C6H6O4 Formula Weight: 142.11 Odour: Odourless Melting Point: 152-155° Beilstein Reference: 120895 Merck Reference: 14,5317 MDL No.: MFCD00006580

Vzorec: C₆H₆O₄
Bod varu: 323 °C (1013 hPa)
Bod tání: 152…155 °C
Hustota: 1,58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006580
CAS číslo: 501-30-4
EINECS: 207-922-4
Merck Index: 14,5317

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR