Tisk…

Oxid platičitý, anhydrous ≥99.95% (metals basis) Pt ≥ 84.4%, Premion®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Platinum dioxide, Adam's type catalyst

40402.01 40402.03 40402.06
40402.01EA 2840 CZK
40402.01 40402.03 40402.06
Oxid platičitý, anhydrous ≥99.95% (metals basis) Pt ≥ 84.4%, Premion®
Oxid platičitý

Purity: 99.95% CAS No.: 1314-15-4 Molecular Formula: O2Pt Formula Weight: 227.09 Physical Form: Powder Density: 10.2 Appearance: Dark brown Odour: Odourless Melting Point: 450° Merck Reference: 14,7527 UN No.: 1479 MDL No.: MFCD00011184

Vzorec: O₂Pt
MW: 227.08
Bod tání: 450
Hustota: 10.2
Storage Temperature: Ambient
MDL Number: MFCD00011184
CAS číslo: 1314-15-4
EINECS: 215-223-0
UN: 1479
ADR: 5.1 II
Merck Index: 14,7527

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR