Iridium(IV) oxide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Iridium dioxide, Iridium(IV) oxide
M-Clarity™ quality level = MQ100
206237-1G
206237-5G
SIAL206237-1GEA
18780
CZK
SIAL206237-1G
SIAL206237-5G
Iridium(IV) oxide, Sigma-Aldrich®
Iridium(IV) oxide
Vzorec:
IrO₂ MW: 224,22 g/mol Bod varu: –27 °C (1013 hPa) Bod tání: –102 °C Hustota: 3,09 g/cm³ (18 °C) |
MDL Number:
MFCD00011065 CAS číslo: 12030-49-8 EINECS: 234-743-9 UN: 1479 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...