Oxid železitý, NanoArc®, industrial
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: 20-40nm APS Powder, S.A. 30-60m2/g
Density: 5.24
Appearance: Red-brown
Odour: Odourless
Melting Point: 1565°
MDL No.: MFCD00011008
NanoArc® industrial iron oxide is a general purpose gamma-phase material suitable for use in catalytic applications such as ammonia synthesis and dehydrogenation reactions. Manufactured by growing nanocrystals rapidly in..
Vzorec:
Fe₂O₃ Bod tání: 1565 °C Hustota: 5,24 Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS číslo: 1309-37-1 EINECS: 215-168-2 Merck Index: 13,04058 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...