Oxid kobaltnato-kobaltitý
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Tricobalt tetraoxide, Cobaltic tetraoxide
A16121.0B
A16121.18
A16121.30
A16121.0BEA
12190
CZK
A16121.0B
A16121.18
A16121.30
Oxid kobaltnato-kobaltitý
Oxid kobaltnato-kobaltitý
CAS No.: 1308-06-1
Molecular Formula: Co3O4
Formula Weight: 240.80
Melting Point: 900° dec.
Merck Reference: 14,2429
UN No.: 3077
MDL No.: MFCD00010939
Vzorec:
Co₃O₄ MW: 240,8 g/mol Bod tání: 900 °C (rozkl.) Hustota: 6,11 Storage Temperature: Ambient |
MDL Number:
MFCD00010939 CAS číslo: 1308-06-1 EINECS: 215-157-2 UN: 3077 ADR: 9,III Merck Index: 14,2429 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...