Oxid kobaltnatý ≥95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Cobalt oxide
23141.14
23141.22
AB204251.0025
23141.14EA
3510
CZK
23141.14
23141.22
ABCRAB204251.0025
Oxid kobaltnatý ≥95%
Oxid kobaltnatý
Purity: 95%
CAS No.: 1307-96-6
Molecular Formula: CoO
Formula Weight: 74.93
Physical Form: Powder
Density: 6.45
Appearance: Black-green
Melting Point: 1935°
Solubility: Soluble in acids or alkalies. Insoluble in water
Merck Reference: 14,2446
UN No.: 2811
MDL No.: MFCD00016031
Pigments, glass Colouring, semiconductors
Vzorec:
CoO Bod tání: 1935 °C Hustota: 6,45 Storage Temperature: Ambient |
MDL Number:
MFCD00016031 CAS číslo: 1307-96-6 EINECS: 215-154-6 UN: 2811 ADR: 6.1,III Merck Index: 14,2446 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...