Tisk…

Cobalt(II) iodide, ultra dry ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Cobaltous iodide

Ampuled under argon

35730.03 35730.06
35730.03EA 4530 CZK
35730.03 35730.06
Cobalt(II) iodide, ultra dry ≥99.999% (metals basis)
Cobalt(II) iodide

Purity: 99.999% CAS No.: 15238-00-3 Molecular Formula: CoI2 Formula Weight: 312.74 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.584 Appearance: White Odour: Odourless Melting Point: 515° vac. Boiling Point: 570° vac. Merck Reference: 14,2443 MDL No.: MFCD00016030

Vzorec: CoI₂
Bod varu: 570 °C (in vacuum)
Hustota: 5,584
Storage Temperature: Ambient
MDL Number: MFCD00016030
CAS číslo: 15238-00-3
EINECS: 239-283-2
Merck Index: 14,2443

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR