Chlorid nikelnatý, ultra dry ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Ampuled under argon
Purity: 99.9%
CAS No.: 7718-54-9
Molecular Formula: NiCl2
Formula Weight: 129.60
Physical Form: Powder, Ampouled under argon
Density: 3.55
Appearance: Orange
Odour: Odourless
Melting Point: 1001°
Merck Reference: 14,6505
UN No.: 3288
MDL No.: MFCD00011142
Vzorec:
Cl₂Ni Bod tání: 1001 °C Hustota: 3,55 Storage Temperature: Ambient |
MDL Number:
MFCD00011142 CAS číslo: 7718-54-9 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III Merck Index: 14,6505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...