Iron(II) chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
429368-10G
429368-1G
SIAL429368-10GEA
14480
CZK
SIAL429368-10G
SIAL429368-1G
Iron(II) chloride, Sigma-Aldrich®
Iron(II) chloride
Vzorec:
Cl₂Fe Bod varu: 2023 °C (1013 hPa) Bod tání: 674 °C Hustota: 2,7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011004 CAS číslo: 7758-94-3 EINECS: 231-843-4 UN: 3260 ADR: 8,III Merck Index: 12,04091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...