Iron(II) chloride, ultra dry ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Iron dichloride, Ferrous chloride
Ampuled under argon
35701.03
35701.09
35701.18
35701.03EA
2580
CZK
35701.03
35701.09
35701.18
Iron(II) chloride, ultra dry ≥99.99% (metals basis)
Iron(II) chloride
Purity: 99.99%
CAS No.: 7758-94-3
Molecular Formula: Cl2Fe
Formula Weight: 126.75
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.160
pH: 2.5
Melting Point: 674°
Boiling Point: 1023°
Merck Reference: 14,4043
UN No.: 3260
MDL No.: MFCD00011004
Vzorec:
Cl₂Fe MW: 126.75 Bod varu: 1023 °C Bod tání: 674 Hustota: 3.160 Storage Temperature: Ambient |
MDL Number:
MFCD00011004 CAS číslo: 7758-94-3 EINECS: 231-843-4 UN: 3260 ADR: 8 III Merck Index: 14,4043 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...