Platinum(II) bromide (≥54,5% Pt) ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
12172.03
12172.06
12172.03EA
4360
CZK
12172.03
12172.06
Platinum(II) bromide (≥54,5% Pt) ≥99.9% (metals basis)
Platinum(II) bromide
Purity: 99.9%
CAS No.: 13455-12-4
Molecular Formula: PtBr2
Formula Weight: 354.91
Physical Form: Crystalline
Melting Point: 250° dec.
MDL No.: MFCD00011180
Vzorec:
Br₂Pt Hustota: 6,652 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011180 CAS číslo: 13455-12-4 EINECS: 236-648-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...