Tisk…

Rhenium dioxide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

342254-1G 342254-5G
SIAL342254-1GEA 18900 CZK
SIAL342254-1G SIAL342254-5G
Rhenium dioxide, Sigma-Aldrich®
Rhenium dioxide
Vzorec: O₂Re
Hustota: 11,4 g/cm³ (25 °C)
MDL Number: MFCD00016305
CAS číslo: 12036-09-8
EINECS: 234-839-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR