Tisk…

Manganese(III) oxide, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Manganese oxide (Mn2O3), Dimanganese trioxide

M-Clarity™ quality level = MQ100

463701-25G 463701-5G
SIAL463701-25GEA 20130 CZK
SIAL463701-25G SIAL463701-5G
Manganese(III) oxide, Sigma-Aldrich®
Manganese(III) oxide
Vzorec: Mn₂O₃
MW: 157,87 g/mol
Bod tání: 1347 °C
Hustota: 4,81 g/cm³ (20 °C)
MDL Number: MFCD00016217
CAS číslo: 1317-34-6
EINECS: 215-264-4
Merck Index: 13,05761

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR