Tisk…

Sodium bitartrate monohydrate ≥98.0% (by titrimetric analysis), purum p.a., Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2R,3R)-2,3-Dihydroxysuccinic acid, sodium salt hydrate

71680-250G 71680-1KG
SIAL71680-250GEA 2100 CZK
SIAL71680-250G SIAL71680-1KG
Sodium bitartrate monohydrate ≥98.0% (by titrimetric analysis), purum p.a., Sigma-Aldrich®
Sodium bitartrate monohydrate

Sodium bitartrate monohydrate (SBTMH) is an organometallic compound having nonlinear optical properties.

Vzorec: C₄H₅NaO₆·H₂O
MW: 190,083 g/mol
MDL Number: MFCD00150038
CAS číslo: 6131-98-2
EINECS: 208-400-9

Objednat

Výsledky specifikačního testu

Assay ≥98.0% (T)
Ca ≤50 mg/kg
Cd ≤50mg/kg
Co ≤50 mg/kg
Cu ≤50 mg/kg
Fe ≤50 mg/kg
K ≤100 mg/kg
Ni ≤50 mg/kg
PB ≤50 mg/kg
Zn ≤50 mg/kg
Chloride (Cl⁻) ≤50 mg/kg
Sulphate (SO₄)²⁻ ≤50 mg/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR