Tisk…

Manganese(II) bromide, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Manganous bromide, Manganese(ii)bromide, Manganese dibromide, Manganese(II) bromide hydrate

M-Clarity™ quality level = MQ100

223646-100G 223646-25G
SIAL223646-100GEA 17180 CZK
SIAL223646-100G SIAL223646-25G
Manganese(II) bromide, Sigma-Aldrich®
Manganese(II) bromide
Vzorec: Br₂Mn
MW: 214,75 g/mol
Bod tání: 698 °C
Hustota: 4,38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011113
CAS číslo: 13446-03-2
EINECS: 236-591-9
Merck Index: 12,05765

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR